Learn More
Medchemexpress LLC Bis(2-bromoethyl) ether | 5414-19-7 | 99.9% | 231.92 | 100 G
SDP
Catalog No. 5000454235
Encompass_Preferred
Bis(2-bromoethyl) ether is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize the ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-004-54235 Supplier Medchemexpress LLC Supplier No. HYW013458100G
Bis(2-bromoethyl) ether is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize the ubiquitin-proteasome system to selectively degrade target proteins