Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis(2-chloroethyl)amine-1,1,2,2-d4 hydrochloride | 352431-06-2 | 98.0% | 182.51 | C4H6D4Cl3N | 25mg
SDP

Catalog No. 5000270009
Encompass_Preferred

Bis(2-chloroethyl)amine-1 1 2 2-d4 (hydrochloride) is the deuterium labeled Bis(2-chloroethyl)amine-1 1 2 2 hydrochloride[1]

Catalog No. 50-002-70009 Supplier Medchemexpress LLC Supplier No. HYY0881S25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis(2-chloroethyl)amine-1 1 2 2-d4 (hydrochloride) is the deuterium labeled Bis(2-chloroethyl)amine-1 1 2 2 hydrochloride[1]

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.