Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-chloroethyl)amine Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. B1210500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1210500G 500 g
B121025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1210500G Supplier TCI America Supplier No. B1210500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 821-48-7
Molecular Formula C4H10Cl3N
Molecular Weight (g/mol) 178.481
MDL Number MFCD00012515
InChI Key YMDZDFSUDFLGMX-UHFFFAOYSA-N
Synonym bis 2-chloroethyl amine hydrochloride, 2,2'-dichlorodiethylamine hydrochloride, 2-chloro-n-2-chloroethyl ethanamine hydrochloride, nor-lost hydrochlorid, nor-hn2, ethanamine, 2-chloro-n-2-chloroethyl-, hydrochloride, n,n-bis 2-chloroethyl amine hydrochloride, di-2-chloroethyl amine hydrochloride, 2,2-dichlorodiethylamine hydrochloride, bis-2-chloroethyl amine hydrochloride
PubChem CID 522769
IUPAC Name 2-chloro-N-(2-chloroethyl)ethanamine;hydrochloride
SMILES C(CCl)NCCCl.Cl

Specifications

Melting Point 213°C
Quantity 500 g
Formula Weight 178.48
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-chloroethyl)amine Hydrochloride
RTECSNumber IA1225000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.