Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(2-chloroethyl) ether

Catalog No. 501859539
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-9539 250 mL
50-185-9538 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-185-9539 Supplier Sigma Aldrich Supplier No. 35660250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(2-chloroethyl) ether
TRUSTED_SUSTAINABILITY

Specifications

CAS 111-44-4
Melting Point -47°C (lit.)
Density 1.22 g/mL (at 25°C (literature))
Boiling Point 65°C to 67°C (15 mmHg)
Molecular Formula C4H8Cl2O
Refractive Index n20/D 1.456 (literature); n20/D 1.457
Linear Formula (ClCH2CH2)2 O
Quantity 250 mL
MDL Number MFCD00000975
Synonym 2,2 -Dichlorodiethyl ether
Molecular Weight (g/mol) 143.01
Percent Purity ≥99.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.