Learn More
Medchemexpress LLC Bis(2-cyanoethyl) diisopropylphosphoramidite | 102690-88-0 | 95.0% | 271.30 | 25 G
SDP
Catalog No. 5000402864
Encompass_Preferred
Bis(2-cyanoethyl) diisopropylphosphoramidite is a phosphorite monomer that can be used in the synthesis of oligonucleotides.
  • It is a phosphorite monomer.
  • It can be used in the synthesis of oligonucleotides.
Catalog No. 50-004-02864 Supplier Medchemexpress LLC Supplier No. 25GHY24126
Bis(2-cyanoethyl) diisopropylphosphoramidite is a phosphorite monomer that can be used in the synthesis of oligonucleotides.
  • It is a phosphorite monomer.
  • It can be used in the synthesis of oligonucleotides.