Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(2-Cyanoethyl)-N,N-Diisopropylphosphoramidite

Catalog No. 7663055G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-630-55G 2 g
76-630-51G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 76-630-55G Supplier Sigma Aldrich Supplier No. 7663055G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Useful phosphorylating reagent used in oligonucleotide synthesis for adding a terminal phosphate group to the 3' or 5' hydroxyl.Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite may be used as a phosphitylating agent during the multi-step synthesis of (-)-1L-chiro-inositol 2,3,5-trisphosphate starting fro mL-quebrachitol.

Specifications

CAS 1218788-80-7
Melting Point 380°C to 385°C
Molecular Formula C39H32CuIOP2
Refractive Index n20/D 1.465
Linear Formula C39H32CuIOP2
MDL Number MFCD23098730
Quantity 2 g
Synonym Iodo[1,1'-(9,9-dimethyl-9H-xanthene-4,5-diyl)bis(1,1-diphenylphosphine-P)]copper(I)
Molecular Weight (g/mol) 769.07 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.