Learn More
Strem, An Ascensus Company CAS# 131890-26-1. 20g. Bis[(2-di-i-propylphosphino)ethyl]amine, min. 97% (10 wt% in tetrahydrofuran). MFCD17018778
SDP
Supplier:  Strem, An Ascensus Company 15730420G
Encompass
CAS# 131890-26-1. 20g. Bis[(2-di-i-propylphosphino)ethyl]amine, min. 97% (10 wt% in tetrahydrofuran). MFCD17018778. Molecular Weight: 305.42. Molecular Formula: C16H37NP2. Color/form: pale yellow to colorless liq. Strem# 15-7304. http://www.strem.com/catalog/v/15-7304/
Catalog No. 50-138-5431