Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis[(2-Dimethylamino)Phenyl]Amine Nickel(II) Chloride

Catalog No. 728551100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-855-1100MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-855-1100MG Supplier Sigma Aldrich Supplier No. 728551100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for:Alkyl-alkyl Kumada coupling of secondary alkyl halidesDirect alkylation of heterocyclic C-H bondsSonogashira coupling of nonactivated alkyl halidesKumada-Corriu-Tamao coupling of nonactivated alkyl halides with aryl and heteroaryl nucleophilesReactant of pincer NN2 ligand leading to selective carbon-carbon bond formation

Specifications

CAS 80028-44-0
Melting Point 17°C to 42°C
Molecular Formula C7H13NO2 -+ HCl
Linear Formula C7H13NO2 -+ HCl
MDL Number MFCD14585012
Quantity 1 g
Synonym Pyrrolidine-3-carboxylic acid ethyl ester hydrochloride
Molecular Weight (g/mol) 179.64 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.