Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) adipate, 99%

Catalog No. AC402460025
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402460025 2.5 kg Glass bottle
AC402462500 250 g Glass bottle
AC402460010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC402460025 Supplier Thermo Scientific Chemicals Supplier No. 402460025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 103-23-1 | C22H42O4 | 370.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-23-1
Molecular Formula C22H42O4
Molecular Weight (g/mol) 370.57
MDL Number MFCD00009496
InChI Key SAOKZLXYCUGLFA-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl adipate, di 2-ethylhexyl adipate, diethylhexyl adipate, deha, plastomoll doa, beha, vestinol oa, bisoflex doa, effomoll doa
PubChem CID 7641
ChEBI CHEBI:34675
IUPAC Name bis(2-ethylhexyl) hexanedioate
SMILES CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -76.0°C
Density 0.9240g/mL
Boiling Point 210.0°C to 218.0°C (5.0 mmHg)
Flash Point 196°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4460 to 1.4480
Quantity 2.5 kg
Specific Gravity 0.924
Solubility Information Solubility in water: insouble. Other solubilities: soluble in aceton,ether,ethanol
Viscosity 13-15 mPa.s (20°C)
Formula Weight 370.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) adipate
Show More Show Less
Hazard Category
EINECSNumber 203-090-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.