Learn More
Medchemexpress LLC Bis(2-ethylhexyl) adipate-d8 | 1214718-98-5 | 100.0% | 378.62 | 1 MG
SDP
Catalog No. 5000453401
Encompass_Preferred
Bis(2-ethylhexyl) adipate-d8 is the deuterium labeled Bis(2-ethylhexyl) adipate. It is intended for research use only and not for sale to patients. This compound is a colorless to light yellow liquid and can serve as a tracer and an internal standard for quantitative analysis. Deuterium substitution is recognized for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • Influences pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-53401 Supplier Medchemexpress LLC Supplier No. HYW009052S1MG
Bis(2-ethylhexyl) adipate-d8 is the deuterium labeled Bis(2-ethylhexyl) adipate. It is intended for research use only and not for sale to patients. This compound is a colorless to light yellow liquid and can serve as a tracer and an internal standard for quantitative analysis. Deuterium substitution is recognized for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • Influences pharmacokinetic and metabolic profiles of drugs