Learn More
Medchemexpress LLC Bis(2-ethylhexyl) adipate-d8 | 1214718-98-5 | 100.0% | 378.62 | 5 MG
SDP
Catalog No. 5000453499
Encompass_Preferred
Bis(2-ethylhexyl) adipate-d8 is the deuterium labeled Bis(2-ethylhexyl) adipate. It is for research use only and not sold to patients. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during the drug development process.
Catalog No. 50-004-53499 Supplier Medchemexpress LLC Supplier No. HYW009052S5MG
Bis(2-ethylhexyl) adipate-d8 is the deuterium labeled Bis(2-ethylhexyl) adipate. It is for research use only and not sold to patients. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during the drug development process.