Learn More
Medchemexpress LLC Bis(2-ethylhexyl) adipate-d8 | 1214718-98-5 | 99.97% | 378.62 | 10 MG
SDP
Catalog No. 5000453144
Encompass_Preferred
Bis(2-ethylhexyl) adipate-d8 is a deuterium labeled version of Bis(2-ethylhexyl) adipate. Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development, and deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-53144 Supplier Medchemexpress LLC Supplier No. HYW009052S10MG
Bis(2-ethylhexyl) adipate-d8 is a deuterium labeled version of Bis(2-ethylhexyl) adipate. Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development, and deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS