Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) hydrogen phosphate, 95%

Catalog No. AC402470050
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass Bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402470050 5 kg Plastic drum
AC402470010 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC402470050 Supplier Thermo Scientific Chemicals Supplier No. 402470050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 298-07-7 | C16H35O4P | 322.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-07-7
Molecular Formula C16H35O4P
Molecular Weight (g/mol) 322.43
MDL Number MFCD00009492
InChI Key SEGLCEQVOFDUPX-UHFFFAOYNA-N
Synonym bis 2-ethylhexyl hydrogen phosphate, bis 2-ethylhexyl phosphate, hdehp, di 2-ethylhexyl phosphate, dehpa extractant, di 2-ethylhexyl phosphoric acid, escaid 100, d 2ehpa, bis 2-ethylhexyl phosphoric acid
PubChem CID 9275
IUPAC Name bis(2-ethylhexyl) hydrogen phosphate
SMILES CCCCC(CC)COP(O)(=O)OCC(CC)CCCC

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -60.0°C
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 48.0°C (12.0 mmHg)
Flash Point 137°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Plastic drum
Refractive Index 1.4400 to 1.4440
Linear Formula (CH3(CH2)3CH(C2H5)CH2O)2POOH
Quantity 5 kg
Beilstein 01,IV,1786
Specific Gravity 0.97
Solubility Information Solubility in water: slightly soluble
Formula Weight 322.42
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) hydrogen phosphate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 206-056-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.