Learn More
Medchemexpress LLC Bis 2-ethylhexyl ma 500mg
SDP
Catalog No. 5000205290
Encompass_Preferred
Bis(2-ethylhexyl) maleate can enhance the water sensitivity of polyvinyl acetate (PVAc) films and improve the tackiness adhesion and wash resistance of polyacrylate[1]
Catalog No. 50-002-05290 Supplier Medchemexpress LLC Supplier No. HYW099735500MG
Bis(2-ethylhexyl) maleate can enhance the water sensitivity of polyvinyl acetate (PVAc) films and improve the tackiness adhesion and wash resistance of polyacrylate[1]