Learn More
Medchemexpress LLC Bis 2-ethylhexyl ma 5g
SDP
Catalog No. 5000205516
Encompass_Preferred
Bis(2-ethylhexyl) maleate can enhance the water sensitivity of polyvinyl acetate (PVAc) films and improve the tackiness adhesion and wash resistance of polyacrylate[1]
Catalog No. 50-002-05516 Supplier Medchemexpress LLC Supplier No. HYW0997355G
Bis(2-ethylhexyl) maleate can enhance the water sensitivity of polyvinyl acetate (PVAc) films and improve the tackiness adhesion and wash resistance of polyacrylate[1]