Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) phosphate, 95%

Extraction of metals

Manufacturer:  Alfa Aesar 1772336

Quantity 500g

 View more versions of this product

Catalog No. AA1772336


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

298-07-7
322.42
1902
Bis(2-ethylhexyl) Phosphate
[CH3(CH2)3CH(C2H5)CH2O]2P(O)(OH)
MFCD00009492
Di(2-ethylhexyl)phosphoric Acid

Specifications

171° (340°F)
?50°C
298-07-7
Liquid
[CH3(CH2)3CH(C2H5)CH2O]2P(O)(OH)
MFCD00009492
Di(2-ethylhexyl)phosphoric Acid
95%
48°C (12mmHg)
500g
0.965
95%
322.42
1902
Bis(2-ethylhexyl) Phosphate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H312

GHS P Statement
P260-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P310-P312-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: DIISOOCTYL ACID PHOSPHATE

RTECSNumber : TB7875000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit