Learn More
Medchemexpress LLC Bis(2-ethylhexyl) phthalate | 117-81-7 | 99.56% | 100 MG
SDP
Catalog No. 5000411053
Encompass_Preferred
DEHP (Bis(2-ethylhexyl) phthalate) is a widely used plasticizer that is orally active. It can produce various toxic effects on organisms, including neurotoxicity, liver toxicity, immunotoxicity, and reproductive toxicity.
  • Inhibits cell viability in mouse GC-1 spg cells
  • Induces autophagy and apoptosis via oxidative stress
  • Impairs fertility in female C3H/N mice
  • Increases body weight and visceral fat tissue
  • Increases expression of PPARα and PPARγ mRNA
Catalog No. 50-004-11053 Supplier Medchemexpress LLC Supplier No. HYB1945100MG
DEHP (Bis(2-ethylhexyl) phthalate) is a widely used plasticizer that is orally active. It can produce various toxic effects on organisms, including neurotoxicity, liver toxicity, immunotoxicity, and reproductive toxicity.
  • Inhibits cell viability in mouse GC-1 spg cells
  • Induces autophagy and apoptosis via oxidative stress
  • Impairs fertility in female C3H/N mice
  • Increases body weight and visceral fat tissue
  • Increases expression of PPARα and PPARγ mRNA