Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl)phthalate-3,4,5,6-d4, MilliporeSigma™ Supelco™

Catalog No. 111015936 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5936 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5936 Supplier MilliporeSigma Supelco Supplier No. 3416425MG
Add to Cart
Add to Cart
Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 93951-87-2
Physical Form Neat
Linear Formula C24D4H34O4
MDL Number MFCD00143759
Melting Point -50°C
Boiling Point 231°C (5 mmHg)
Synonym DEHP-3,4,5,6-d4; Deuterated DEHP; Deuterated bis(2-ethylhexyl)phthalate; Dioctyl phthalate-3,4,5,6-d4; Phthalic acid-3,4,5,6-d4 bis(2-ethylhexyl ester)
Type Analytical Standard
Quantity 25 mg
Formula Weight 394.58
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.