Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis(2-ethylhexyl) phthalate (deuterated) | 352431-42-6 | 428.79 | C24D38O4 | 10 MG
SDP

Catalog No. 5000297639
Encompass_Preferred

DEHP-d38 is the deuterium-labeled form of bis(2-ethylhexyl) phthalate used as an analytical standard and tracer in mass spectrometry and quantitative studies. Supplied in a small research-scale package, it facilitates accurate quantification and recovery experiments where isotopic labeling improves analytical precision.

  • Deuterium-labeled internal standard for mass spectrometry.
  • Molecular formula C24D38O4, molecular weight 428.79, CAS 352431-42-6.
  • Liquid, colorless to light yellow appearance.
  • Stable under recommended storage: -20°C for long-term; -80°C in solvent for short-term.
  • Available in small research quantities, including 10 mg packages.

Catalog No. 50-002-97639 Supplier Medchemexpress LLC Supplier No. HYB1945S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DEHP-d38 is the deuterium-labeled form of bis(2-ethylhexyl) phthalate used as an analytical standard and tracer in mass spectrometry and quantitative studies. Supplied in a small research-scale package, it facilitates accurate quantification and recovery experiments where isotopic labeling improves analytical precision.

  • Deuterium-labeled internal standard for mass spectrometry.
  • Molecular formula C24D38O4, molecular weight 428.79, CAS 352431-42-6.
  • Liquid, colorless to light yellow appearance.
  • Stable under recommended storage: -20°C for long-term; -80°C in solvent for short-term.
  • Available in small research quantities, including 10 mg packages.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.