Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(2-ethylhexyl) sebacate

Catalog No. 2908311L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
3 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-083-11L 1 L
29-083-13L 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-083-11L Supplier Sigma Aldrich Supplier No. 2908311L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(2-ethylhexyl) sebacate, or dioctyl sebacate (DOS), is a non-polar plasticizer that has two ester groups that bond with cationic compounds. It is mainly used in the formation of thin plasticized polyvinyl chloride (PVC) based membranes.DOS can be used as an aerosol material that can be used to investigate the filtration efficiency of polyvinylidene/silicon oxide (PVDF/SiO2) based fibrous membranes. It may be blended with PVC to form membranes that can be deposited on silver (Ag) based composites which can be used in the fabrication of ion-selective electrodes for chemical analysis. DOS may also be used in combination with PVC, chloride ionophore III, and tridodecylmethylammonium chloride for the development of chloride sensing films.

Specifications

CAS 122-62-3
Density 0.914 g/mL (at 25°C)
Boiling Point 212°C (1 mmHg)
Molecular Formula C26H50O4
Refractive Index n20/D 1.450 (literature)
Linear Formula [-(CH2)4 CO2CH2CH(C2H5)(CH2)3 CH3]2
Quantity 1 L
MDL Number MFCD00009497
Synonym Di(2-ethylhexyl) sebacate; Sebacic acid di(2-ethylhexyl) ester; Dioctyl sebacate
Molecular Weight (g/mol) 426.67
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.