Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) sulfoxide, Thermo Scientific™

Catalog No. AAL1129306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1129306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1129306 Supplier Thermo Scientific Chemicals Supplier No. L1129306

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82374-34-3
Molecular Formula C16H34OS
Molecular Weight (g/mol) 274.51
MDL Number MFCD00040086
InChI Key PGRIEACWYQKVQY-UHFFFAOYNA-N
Synonym bis 2-ethylhexyl sulfoxide, 3-2-ethylhexylsulfinylmethyl heptane, acmc-20alo3, 3-2-ethylhexanesulfinyl methyl heptane, heptane, 3,3'-sulfinylbis methylene bis
PubChem CID 3813850
IUPAC Name 3-[(2-ethylhexanesulfinyl)methyl]heptane
SMILES CCCCC(CC)CS(=O)CC(CC)CCCC

Specifications

Quantity 5 g
Beilstein 4973978
Formula Weight 274.51
Chemical Name or Material Bis(2-ethylhexyl) sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.