Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) terephthalate, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015037 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5037 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5037 Supplier MilliporeSigma Supelco Supplier No. 492341ML
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 6422-86-2
Vapor Pressure 1 mmHg (217°C)
Density 0.986 g/mL (at 25°C)
Flash Point 212°C - Closed Cup
Linear Formula C6H4-1,4-[CO2 CH2 CH(C2H5)(CH2)3 CH3]2
MDL Number MFCD00072256
Melting Point -48°C
Boiling Point 400°C
Synonym Dioctyl terephthalate; Bis(2-ethylhexyl) terephthalate
Type Analytical Standard
Quantity 1 mL
Formula Weight 390.56
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.