Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) terephthalate, 97%, Thermo Scientific™

Catalog No. AC402490100
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC402490100 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC402490100 Supplier Thermo Scientific Chemicals Supplier No. 402490100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6422-86-2
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.55
InChI Key RWPICVVBGZBXNA-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl terephthalate, kodaflex dotp, di-2-ethylhexyl terephthalate, 1,4-benzenedicarboxylic acid, bis 2-ethylhexyl ester, terephthalic acid, bis 2-ethylhexyl ester, eastman 168, ccris 7052, di 2-ethylhexyl terephthalate, bis 2-ethylhexyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-bis 2-ethylhexyl ester
PubChem CID 22932
IUPAC Name bis(2-ethylhexyl) benzene-1,4-dicarboxylate
SMILES CCCCC(CC)COC(=O)C1=CC=C(C=C1)C(=O)OCC(CC)CCCC

Specifications

Melting Point -67.0°C
Density 0.9840g/mL
Boiling Point 400.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Refractive Index 1.4890 to 1.4910
Quantity 10 kg
Beilstein 09,IV,3306
Specific Gravity 0.984
Solubility Information Solubility in water: insoluble.
Formula Weight 390.55
Percent Purity 97%
Chemical Name or Material Bis(2-ethylhexyl) terephthalate
Show More Show Less
EINECSNumber 229-176-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.