Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methane, 94%, Thermo Scientific™

Catalog No. AAL1332330
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1332330 250 g
AAL1332318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1332330 Supplier Thermo Scientific Chemicals Supplier No. L1332330

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-62-3
Molecular Formula C29H40O2
Molecular Weight (g/mol) 420.64
MDL Number MFCD00151797
InChI Key PHXLONCQBNATSL-UHFFFAOYSA-N
Synonym 2,2'-methylenebis 6-1-methylcyclohexyl-p-cresol, ionox wsp, nonox wsp, bisalkofen mtsp, unii-7tir91vdi9, 7tir91vdi9, p-cresol, 2,2'-methylenebis 6-1-methylcyclohexyl, 2,2'-methylenebis 4-methyl-6-1-methylcyclohexyl phenol
PubChem CID 6486
IUPAC Name 2-[[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methyl]-4-methyl-6-(1-methylcyclohexyl)phenol
SMILES CC1=CC(=C(O)C(CC2=C(O)C(=CC(C)=C2)C2(C)CCCCC2)=C1)C1(C)CCCCC1

Specifications

Melting Point 130°C to 135°C
Odor Odorless
Quantity 250 g
Beilstein 2313064
Formula Weight 420.64
Percent Purity 94%
Chemical Name or Material Bis[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methane
EINECSNumber 201-044-5
RTECSNumber SM0400000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.