Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-hydroxyethyl) Disulfide, MP Biomedicals

Catalog No. ICN212869 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212869 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212869 Supplier MP Biomedicals, Inc Supplier No. 0521286910

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3570-55-6
Molecular Formula C4H10S3
Molecular Weight (g/mol) 154.30
MDL Number MFCD00004893
InChI Key KSJBMDCFYZKAFH-UHFFFAOYSA-N
Synonym 2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi
PubChem CID 77117
IUPAC Name 2-[(2-sulfanylethyl)sulfanyl]ethane-1-thiol
SMILES SCCSCCS

Specifications

Quantity 10 g
Formula Weight 154.25
Chemical Name or Material Bis(2-hydroxyethyl)Disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.