Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-hydroxyphenyl)methane, MP Biomedicals

Catalog No. ICN201363 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN201363 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN201363 Supplier MP Biomedicals, Inc Supplier No. 0520136301

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2474-07-9
Molecular Formula C10H14BrNO
Molecular Weight (g/mol) 244.13
InChI Key MOVOYJFCKMYLHQ-UHFFFAOYSA-N
Synonym 2-4-bromophenoxy-n,n-dimethylethanamine, 2-4-bromophenoxy-n,n-dimethylethylamine, 2-4-bromophenoxy ethyl dimethylamine, 4-2-n,n-dimethylethoxy phenylbromide, 4-2-n,n-dimethylaminoethoxy phenylbromide, 2-4-bromo-phenoxy-ethyl-dimethyl-amine, ethanamine, 2-4-bromophenoxy-n,n-dimethyl, n-2-4-bromophenoxy ethyl-n,n-dimethylamine, 2-4-bromo-phenoxy-ethyl-dimethyl, 4-2-dimethylaminoethoxy bromobenzene
PubChem CID 213877
IUPAC Name 2-(4-bromophenoxy)-N,N-dimethylethanamine
SMILES CN(C)CCOC1=CC=C(C=C1)Br

Specifications

Quantity 1 g
Formula Weight 200.24
Chemical Name or Material Bis(2-hydroxyphenyl)methane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.