Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(2-hydroxypropyl)amine

Catalog No. 501846475
Encompass
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-184-6475 250 g
14-960-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-184-6475 Supplier Sigma Aldrich Supplier No. 14960250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(2-hydroxypropyl)amine
TRUSTED_SUSTAINABILITY

Specifications

CAS 110-97-4
Melting Point 42°C to 45°C (lit.)
Density 1.004 g/mL (at 25°C (literature))
Boiling Point 249°C to 250°C (745 mmHg)
Molecular Formula C6H15NO2
Linear Formula NH[CH2CH(OH)CH3]2
MDL Number MFCD00004531
Quantity 250 g
Synonym 1,1 -Iminodi-2-propanol; Diisopropanolamine
Molecular Weight (g/mol) 133.19
Percent Purity ≥98.0% (T)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.