Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis(2-methyl-3-furyl)disulfide | 28588-75-2 | 98.37% | 226.31 | 1 ML
SDP

Catalog No. 5000452059
Encompass_Preferred

Bis(2-methyl-3-furyl)disulfide, also known as compound (2), is a flavoring compound designed to enhance the natural, fresh, and milk-rich feeling of milk-related products. It is for research use only and is not intended for patient use. This compound appears as a liquid with a density of 1.211 g/cm³ and ranges in color from light yellow to brown.

  • Enhances natural, fresh, and milk-rich feeling in milk-related products.
  • For research use only.
  • Appears as a liquid.
  • Density: 1.211 g/cm³.
  • Color: light yellow to brown.

Catalog No. 50-004-52059 Supplier Medchemexpress LLC Supplier No. HYW00970810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis(2-methyl-3-furyl)disulfide, also known as compound (2), is a flavoring compound designed to enhance the natural, fresh, and milk-rich feeling of milk-related products. It is for research use only and is not intended for patient use. This compound appears as a liquid with a density of 1.211 g/cm³ and ranges in color from light yellow to brown.

  • Enhances natural, fresh, and milk-rich feeling in milk-related products.
  • For research use only.
  • Appears as a liquid.
  • Density: 1.211 g/cm³.
  • Color: light yellow to brown.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.