Learn More
Medchemexpress LLC Bis(2-methyl-3-furyl)disulfide | 28588-75-2 | 98.37% | 226.31 | 5 G
SDP
Catalog No. 5000451589
Encompass_Preferred
Bis(2-methyl-3-furyl)disulfide is a chemical compound designed to function as a flavoring agent. It is intended to enhance natural, fresh, and milk-rich sensations in milk-related products. This product is strictly for research use only and is not sold to patients.
  • Enhances natural, fresh, and milk-rich feelings in milk-related products.
  • Provided as a liquid with a density of 1.211 g/cm³.
  • Appears light yellow to brown in color.
  • Stable for up to 3 years in pure form at -20°C.
  • Soluble in DMSO for in vitro applications.
  • Offers specific in vivo dissolution protocols for research flexibility.
Catalog No. 50-004-51589 Supplier Medchemexpress LLC Supplier No. HYW0097085G
Bis(2-methyl-3-furyl)disulfide is a chemical compound designed to function as a flavoring agent. It is intended to enhance natural, fresh, and milk-rich sensations in milk-related products. This product is strictly for research use only and is not sold to patients.
  • Enhances natural, fresh, and milk-rich feelings in milk-related products.
  • Provided as a liquid with a density of 1.211 g/cm³.
  • Appears light yellow to brown in color.
  • Stable for up to 3 years in pure form at -20°C.
  • Soluble in DMSO for in vitro applications.
  • Offers specific in vivo dissolution protocols for research flexibility.