Learn More
Strem, An Ascensus Company BIS(2-METHYLALLYL)(1 5-C-0.25G
SDP
Catalog No. 5090111734
Encompass
Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium (II), min. 97%; Size: 250mg; [12289-94-0]; (C4H7)2(C8H12)Ru; Mol Wt:319.45; Color & Form: tan to brown pwdr.
Catalog No. 50-901-11734 Supplier Strem, An Ascensus Company Supplier No. 440046250MG
Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium (II), min. 97%; Size: 250mg; [12289-94-0]; (C4H7)2(C8H12)Ru; Mol Wt:319.45; Color & Form: tan to brown pwdr.