Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-methylallyl) carbonate, 97%, Thermo Scientific™

Catalog No. AC170580050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170580050 5 g Glass bottle
AC170580250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170580050 Supplier Thermo Scientific Chemicals Supplier No. 170580050
Add to Cart

Chemical Identifiers

CAS 64057-79-0
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00008598
InChI Key SKCUYCAPQSWGBP-UHFFFAOYSA-N
Synonym dimethallyl carbonate, bis 2-methylallyl carbonate, bis methallyl carbonate, bis 2-methyl-2-propenyl carbonate, 2-propen-1-ol, 2-methyl-, carbonate 2:1, 2-methyl-2-propen-1-ol carbonate 2:1, 2-propen-1-ol, 2-methyl-, 1,1'-carbonate, carbonic acid, bis 2-methylallyl ester, bis 2-methylprop-2-en-1-yl carbonate, acmc-1biju
PubChem CID 47083
IUPAC Name bis(2-methylprop-2-enyl) carbonate
SMILES CC(=C)COC(=O)OCC(C)=C

Specifications

Boiling Point 199.0°C to 202.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4371
Linear Formula [H2C=C(CH3)CH2O]2CO
Quantity 5 g
Formula Weight 170.21
Percent Purity 97%
Chemical Name or Material Bis(2-methylallyl) carbonate
EINECSNumber 264-645-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.