Learn More
Strem, An Ascensus Company CAS# 12081-18-4. 500mg. Bis(2-methylallyl)palladium chloride dimer, min. 98%. MFCD02684573
SDP
Supplier:  Strem, An Ascensus Company 460240500MG
Encompass
CAS# 12081-18-4. 500mg. Bis(2-methylallyl)palladium chloride dimer, min. 98%. MFCD02684573. Molecular Weight: 393.94. Molecular Formula: [C4H7PdCl]2. Color/form: yellow pwdr. Strem# 46-0240. http://www.strem.com/catalog/v/46-0240/
Catalog No. 50-901-12295