Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis[2-(Trimethylsilyl)Ethyl] N,N-Diisopropylphosphoramidite

Catalog No. 76081125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-081-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 76-081-125G Supplier Sigma Aldrich Supplier No. 76081125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phosphitylation agent used in the preparation of oligonucleotides

Specifications

CAS 1245823-51-1
Melting Point 118°C to 125°C
Molecular Formula C21H19NO3
Refractive Index n20/D 1.4517
Linear Formula C21H19NO3
MDL Number MFCD22380737
Quantity 25 g
Synonym DBCO-Acid
Molecular Weight (g/mol) 333.38 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.