Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[2-(trimethylsilyloxy)ethyl] Ether 98.0+%, TCI America™
SDP

Catalog No. B337825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B337825G 25 g
B33785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B337825G Supplier TCI America Supplier No. B337825G

Chemical Identifiers

CAS 16654-74-3
Molecular Formula C10H26O3Si2
Molecular Weight (g/mol) 250.485
InChI Key VZCNNAQIFXLAEZ-UHFFFAOYSA-N
Synonym Diethylene Glycol Bis(trimethylsilyl Ether)
PubChem CID 552860
IUPAC Name trimethyl-[2-(2-trimethylsilyloxyethoxy)ethoxy]silane
SMILES C[Si](C)(C)OCCOCCO[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 87°C
Quantity 25 g
Formula Weight 250.49
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis[2-(trimethylsilyloxy)ethyl] Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.