Learn More
Cayman Chemical Bis-(2-chloroisopropyl) ether, Purity: ≥98%, CAS: 108-60-1, 50 mg
SDP
Catalog No. 502984980
Encompass_Preferred
A byproduct in the synthesis of ethylene and propylene glycol; has been found in municipal drinking water in the United States and waterways in the Netherlands where it is considered a POP; mutagenic in the S. typhimurium strains TA 1535 and TA 100
Catalog No. 50-298-4980 Supplier Cayman Chemical Supplier No. 2068350 MG
A byproduct in the synthesis of ethylene and propylene glycol; has been found in municipal drinking water in the United States and waterways in the Netherlands where it is considered a POP; mutagenic in the S. typhimurium strains TA 1535 and TA 100