Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(3,5-Bis(Trifluoromethyl)Phenyl)(2',6'-Bis(Dimethylamino)-3,6-Dimethoxybiphenyl-2-Yl)Phosphine

Catalog No. 900277250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-027-7250MG 1 g
90-027-71G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-027-7250MG Supplier Sigma Aldrich Supplier No. 900277250MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(3,5-Bis(Trifluoromethyl)Phenyl)(2',6'-Bis(Dimethylamino)-3,6-Dimethoxybiphenyl-2-Yl)Phosphine

Specifications

CAS 2197989-24-3
Melting Point 160.3°C
Molecular Formula C34H29F12N2O2P
Linear Formula C34H29F12N2O2P
Quantity 1 g
Synonym 2'-(Bis(3,5-bis(trifluoromethyl)phenyl)phosphanyl)-3',6'-dimethoxy-N ,N ,N ,N -tetramethyl-[1,1'-biphenyl]-2,6-diamine; 2'-(Bis(3,5-bis(trifluoromethyl)phenyl)phosphanyl)-3',6'-dimethoxyN2,N2,N6,N6 -tetramethyl-[1,1'-biphenyl]-2,6-diamine; 2-{Bis[3,5-bis(trifluoromethyl)phenyl]phosphino}-3,6-dimethoxy -2',6'-bis(dimethylamino)-1,1'-biphenyl
Molecular Weight (g/mol) 756.56 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.