Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis(3,5-di-tert-butylphenyl) phosphine oxide | 325773-65-7 | MFCD08458886 | 95.0% | 426.61 g/mol | C28H43OP | 100 MG
SDP

Catalog No. 5000209880
Encompass_Preferred

Bis(3,5-di-tert-butylphenyl) phosphine oxide is an organophosphorus compound used as a drug intermediate in organic synthesis. It is supplied as a solid reagent for laboratory research and small-scale synthetic applications.

  • Drug intermediate for synthesis of active compounds.
  • Empirical formula C28H43OP and molecular weight 426.61 g/mol.
  • CAS number 325773-65-7 for unambiguous identification.
  • Available in small research sizes, such as 100 mg, for laboratory use.
  • Reported purity varies by supplier (commonly reported around 95%).

Catalog No. 50-002-09880 Supplier Medchemexpress LLC Supplier No. HYW047043100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Bis(3,5-di-tert-butylphenyl) phosphine oxide is an organophosphorus compound used as a drug intermediate in organic synthesis. It is supplied as a solid reagent for laboratory research and small-scale synthetic applications.

  • Drug intermediate for synthesis of active compounds.
  • Empirical formula C28H43OP and molecular weight 426.61 g/mol.
  • CAS number 325773-65-7 for unambiguous identification.
  • Available in small research sizes, such as 100 mg, for laboratory use.
  • Reported purity varies by supplier (commonly reported around 95%).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.