Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis(3,5-di-tert-butylphenyl) phosphine oxide | 325773-65-7 | MFCD08458886 | 97.0% | 426.61 g/mol | C28H43OP | 500 MG
SDP

Catalog No. 5000210095
Encompass_Preferred

Bis(3,5-di-tert-butylphenyl) phosphine oxide is a phosphine oxide used as a drug intermediate and research reagent in organic synthesis. It is typically supplied in small lab-scale quantities for use as a building block in medicinal chemistry and specialty materials synthesis.

  • Cas number 325773-65-7.
  • Molecular formula C28H43OP.
  • Molecular weight about 426.61 g/mol.
  • Used as a drug intermediate and research chemistry reagent.
  • Available in small pack sizes (examples: 100 mg, 250 mg, 500 mg, 1 g).

Catalog No. 50-002-10095 Supplier Medchemexpress LLC Supplier No. HYW047043500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Bis(3,5-di-tert-butylphenyl) phosphine oxide is a phosphine oxide used as a drug intermediate and research reagent in organic synthesis. It is typically supplied in small lab-scale quantities for use as a building block in medicinal chemistry and specialty materials synthesis.

  • Cas number 325773-65-7.
  • Molecular formula C28H43OP.
  • Molecular weight about 426.61 g/mol.
  • Used as a drug intermediate and research chemistry reagent.
  • Available in small pack sizes (examples: 100 mg, 250 mg, 500 mg, 1 g).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.