Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis(3,5-di-tert-butylphenyl)phosphine oxide | 325773-65-7 | MFCD08458886 | 97.0% | 426.62 g/mol | C28H43OP | 1 G
SDP

Catalog No. 5000210349
Encompass_Preferred

Bis(3,5-di-tert-butylphenyl)phosphine oxide is a phosphine-oxide drug intermediate used for the synthesis of various active compounds. It is supplied as a powder and intended for research use only.

  • Used as a chemical intermediate for synthesis
  • Physical form: powder
  • Molecular formula: C28H43OP
  • Molecular weight: 426.62 g/mol
  • Typical purity: 97.0%
  • Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month
  • Hazard summary: harmful if swallowed; causes skin and serious eye irritation; may cause respiratory irritation

Catalog No. 50-002-10349 Supplier Medchemexpress LLC Supplier No. HYW0470431G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Bis(3,5-di-tert-butylphenyl)phosphine oxide is a phosphine-oxide drug intermediate used for the synthesis of various active compounds. It is supplied as a powder and intended for research use only.

  • Used as a chemical intermediate for synthesis
  • Physical form: powder
  • Molecular formula: C28H43OP
  • Molecular weight: 426.62 g/mol
  • Typical purity: 97.0%
  • Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month
  • Hazard summary: harmful if swallowed; causes skin and serious eye irritation; may cause respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.