Learn More
Strem, An Ascensus Company CAS# 142421-57-6. 500mg. Bis(3,5-di(trifluoromethyl)phenyl)chlorophosphine, min. 98%. MFCD01630852
SDP
Catalog No. 5090109760
Encompass
CAS# 142421-57-6. 500mg. Bis(3,5-di(trifluoromethyl)phenyl)chlorophosphine, min. 98%. MFCD01630852. Molecular Weight: 492.63. Molecular Formula: C16H6ClF12P. Color/form: colorless to white liquid to melted solid Strem# 15-0114. http://www.strem.com/catalog/v/15-0114/
Catalog No. 50-901-09760 Supplier Strem, An Ascensus Company Supplier No. 150114500MG
CAS# 142421-57-6. 500mg. Bis(3,5-di(trifluoromethyl)phenyl)chlorophosphine, min. 98%. MFCD01630852. Molecular Weight: 492.63. Molecular Formula: C16H6ClF12P. Color/form: colorless to white liquid to melted solid Strem# 15-0114. http://www.strem.com/catalog/v/15-0114/