Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(3,5-Dimethyl-4-Methoxyphenyl)Chlorophosphine

Catalog No. 7103851G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-038-51G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 71-038-51G Supplier Sigma Aldrich Supplier No. 7103851G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Asymmetric organocatalytic electrophilic phosphinationAsymmetric hydroformulation using Taddol-based chiral phosphine-phosphite ligandsSynthesis of ferrocene-based chiral diphosphinesAlcoholysis of phosphane-boranesAsymmetric hydrogenations of bifep-type biferrocenes

Specifications

CAS 32627-01-3
Melting Point 85°C to 87°C
Molecular Formula C12H15O2Rh
Refractive Index n20/D 1.597
Linear Formula C12H15O2Rh
MDL Number MFCD00058847
Quantity 100 mg
Molecular Weight (g/mol) 294.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.