Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(3,5-Dimethylphenyl)Phosphine

Catalog No. 695157500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-515-7500MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 69-515-7500MG Supplier Sigma Aldrich Supplier No. 695157500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(3,5-dimethylphenyl)phosphine can be used as a reactant to prepare: Iron(II) chiral diimine diphosphine complexes as catalysts for the asymmetric transfer hydrogenation of ketones. The complex of iridium with biphenol phosphite-phosphine bidentate ligand as an asymmetric catalyst for the hydrogenation of N-arylimines. Chiral phosphines with excellent stereoselectivity by 1, 4 addition reaction with +a,+a-unsaturated aldehydes using a palladium catalyst.Product of Kanata Chemical Technologies, Inc.

Specifications

CAS 12132-87-5
Melting Point 184°C to 185°C
Molecular Formula C14H10O4Ru2
Linear Formula C14H10O4Ru2
Refractive Index n20/D 1.599
MDL Number MFCD00070447
Quantity 1 g
Synonym Bis(cyclopentadienylruthenium dicarbonyl); Di-mu-carbonyldicarbonylbis(eta5-2,4-cyclopentadien-1-yl)di-ruthenium; Tetracarbonyldicyclopentadienyldi-ruthenium
Molecular Weight (g/mol) 444.37 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.