Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(3,5-dimethylphenyl)phosphine oxide, 98%, Thermo Scientific™

Catalog No. AAH6132006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6132006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6132006 Supplier Thermo Scientific Chemicals Supplier No. H6132006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 187344-92-9
Molecular Formula C16H18OP+
Molecular Weight (g/mol) 257.293
MDL Number MFCD08458884
InChI Key LMXRTXPFJNGAAX-UHFFFAOYSA-N
Synonym bis 3,5-dimethylphenyl phosphine oxide, xyl 2p o h, phosphine oxide,bis 3,5-dimethylphenyl, phosphine oxide, bis 3,5-dimethylphenyl, 1-3,5-dimethylphenyl phosphoryl-3,5-dimethylbenzene, 1-3,5-dimethylphenylphosphoroso-3,5-dimethylbenzene, ksc496c0h, bis 3,5-dimethylphenyl phosphi, 3-4-nitrophenyl-beta-alanine, awaysumoanjulo-uhfffaoysa-n
PubChem CID 12022244
IUPAC Name bis(3,5-dimethylphenyl)-oxophosphanium
SMILES CC1=CC(=CC(=C1)[P+](=O)C2=CC(=CC(=C2)C)C)C

Specifications

Melting Point 82°C to 84°C
Color White
Quantity 5 g
Physical Form Powder
Formula Weight 258.3
Percent Purity 98%
Chemical Name or Material Bis(3,5-dimethylphenyl)phosphine oxide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.