Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(3-biphenylyl)amine 98.0+%, TCI America™
SDP

Catalog No. B5048200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5048200MG 200 mg
B50481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5048200MG Supplier TCI America Supplier No. B5048200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 169224-65-1
Molecular Formula C24H19N
Molecular Weight (g/mol) 321.42
MDL Number MFCD28129440
InChI Key LXOCTSJQHHCASE-UHFFFAOYSA-N
Synonym 3,3′C-Iminobis(biphenyl), Di([1,1′C-biphenyl]-3-yl)amine
PubChem CID 19064202
IUPAC Name N-{[1,1'-biphenyl]-3-yl}-[1,1'-biphenyl]-3-amine
SMILES N(C1=CC=CC(=C1)C1=CC=CC=C1)C1=CC=CC(=C1)C1=CC=CC=C1

Specifications

Melting Point 92°C
Quantity 200 mg
Formula Weight 321.42
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Bis(3-biphenylyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.