Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(3-bromophenyl)acetylene 97.0+%, TCI America™
SDP

Catalog No. B48531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B48531G Supplier TCI America Supplier No. B48531G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 153404-60-5
Molecular Formula C14H8Br2
Molecular Weight (g/mol) 336.03
MDL Number MFCD11111154
InChI Key WSXUXTVJTIPBAF-UHFFFAOYSA-N
PubChem CID 12022272
IUPAC Name 1-bromo-3-[2-(3-bromophenyl)ethynyl]benzene
SMILES BrC1=CC=CC(=C1)C#CC1=CC(Br)=CC=C1

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 1 g
Formula Weight 336.03
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(3-bromophenyl)acetylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.