Learn More
Medchemexpress LLC ((bis(3-chloro-4-methylphenyl)boraneyl)oxy)(3-hydroxypyridin-2-yl)methanone | 872044-70-7 | MFCD18251581 | 99.0% | 400.06 g/mol | C20H16BCl2NO3 | 5 MG
SDP
Catalog No. 5000263903
Encompass_Preferred
AN0128 is a boron-containing antibacterial and anti-inflammatory research compound with demonstrated in vitro activity against several Gram-positive bacteria. It is intended for use in preclinical research on periodontal and cutaneous diseases and is supplied as a solid with high purity; handle protected from light.
Catalog No. 50-002-63903 Supplier Medchemexpress LLC Supplier No. HY109795MG
AN0128 is a boron-containing antibacterial and anti-inflammatory research compound with demonstrated in vitro activity against several Gram-positive bacteria. It is intended for use in preclinical research on periodontal and cutaneous diseases and is supplied as a solid with high purity; handle protected from light.