Learn More
Medchemexpress LLC Bis(3-sulfo-N-succinimidyl) suberate disodium salt | 127634-19-9 | MFCD00054982 | 98.0% | 572.43 | C16H18N2Na2O14S2 | 10 MG
SDP
Catalog No. 5000202214
Encompass_Preferred
BS3 Crosslinker disodium is a water-soluble, non-cleavable, homobifunctional amine-reactive crosslinker used to form stable amide bonds between primary amines. It is commonly used for antibody-drug conjugate synthesis, protein crosslinking, and bioconjugation workflows. CAS 127634-19-9; molecular weight 572.43; typical purity 98%.
  • Water-soluble homobifunctional crosslinker for amine-to-amine coupling.
  • Non-cleavable sulfo-NHS ester chemistry produces stable amide bonds.
  • Membrane-impermeable, suitable for surface and extracellular protein crosslinking.
  • Widely used in antibody-drug conjugate design and general bioconjugation.
  • Provided as a solid powder with high stated purity for research use only.
  • Available in small-scale package sizes for laboratory experiments.
Catalog No. 50-002-02214 Supplier Medchemexpress LLC Supplier No. HY124329A10MG
BS3 Crosslinker disodium is a water-soluble, non-cleavable, homobifunctional amine-reactive crosslinker used to form stable amide bonds between primary amines. It is commonly used for antibody-drug conjugate synthesis, protein crosslinking, and bioconjugation workflows. CAS 127634-19-9; molecular weight 572.43; typical purity 98%.
  • Water-soluble homobifunctional crosslinker for amine-to-amine coupling.
  • Non-cleavable sulfo-NHS ester chemistry produces stable amide bonds.
  • Membrane-impermeable, suitable for surface and extracellular protein crosslinking.
  • Widely used in antibody-drug conjugate design and general bioconjugation.
  • Provided as a solid powder with high stated purity for research use only.
  • Available in small-scale package sizes for laboratory experiments.