Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis[3-(Triethoxysilyl)Propyl] Tetrasulfide

Catalog No. 1520050ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-200-50ML 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-200-50ML Supplier Sigma Aldrich Supplier No. 1520050ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis[3-(triethoxysilyl)propyl] tetrasulfide (TESPTS) is generally used as a crosslinking agent and reinforcing filler in the manufacturing of rubber. TESPT film provides corrosion protection to the metallic substrates. It is also used as a precursor in the synthesis of mesoporous organosilicas.

Specifications

CAS 56-93-9
Density 1.072 g/mL at 20°C
Molecular Formula C10H16ClN
Refractive Index n20/D 1.49
Linear Formula C6H5CH2N(Cl)(CH3)3
MDL Number MFCD00011782
Quantity 1 L
Synonym TESPTS; 3,3'-Tetrathiobis(propyl-triethoxysilane)
Molecular Weight (g/mol) 185.69 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.