Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[4-(3-aminophenoxy)phenyl] Sulfone 98.0+%, TCI America™
SDP

Catalog No. B168125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B168125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B168125G Supplier TCI America Supplier No. B168125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 30203-11-3
Molecular Formula C24H20N2O4S
Molecular Weight (g/mol) 432.49
MDL Number MFCD00054738
InChI Key WCXGOVYROJJXHA-UHFFFAOYSA-N
Synonym 3,3'-sulfonylbis 4,1-phenylene bis oxy dianiline, bis 4-3-aminophenoxy phenyl sulfone, 4,4'-bis 3-aminophenoxy diphenyl sulfone, benzenamine, 3,3'-sulfonylbis 4,1-phenyleneoxy bis, 4,4-bis 3-aminophenoxy diphenyl sulfone, 3-4-4-3-aminophenoxy benzenesulfonyl phenoxy aniline, 4-3-aminophenoxy-1-4-3-aminophenoxy phenyl sulfonyl benzene, acmc-209hds, 3,3'-sulfonylbis 4,1-phenyleneoxy dianiline
PubChem CID 121657
IUPAC Name 3-{4-[4-(3-aminophenoxy)benzenesulfonyl]phenoxy}aniline
SMILES NC1=CC(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(OC3=CC(N)=CC=C3)C=C2)=CC=C1

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 25 g
Formula Weight 432.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis[4-(3-aminophenoxy)phenyl] Sulfone
EINECSNumber (3)-2174
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.