Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[4-(4-aminophenoxy)phenyl] Sulfone 98.0+%, TCI America™
SDP

Catalog No. B168225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B168225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B168225G Supplier TCI America Supplier No. B168225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13080-89-2
Molecular Formula C24H20N2O4S
Molecular Weight (g/mol) 432.49
MDL Number MFCD00039155
InChI Key UTDAGHZGKXPRQI-UHFFFAOYSA-N
PubChem CID 25689
IUPAC Name 4-{4-[4-(4-aminophenoxy)benzenesulfonyl]phenoxy}aniline
SMILES NC1=CC=C(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(OC3=CC=C(N)C=C3)C=C2)C=C1

Specifications

Melting Point 195°C
Quantity 25 g
UN Number 2811
Formula Weight 432.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis[4-(4-aminophenoxy)phenyl] Sulfone
EINECSNumber (3)-4190
RTECSNumber BY8955000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.